Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFPSRRKAA QLPWEDGRSG LLSGGLPRKC SVFHLFVACL SLGFFSLLWL QLSCSGDVAR AVRGQGQETS GPPRACPPEP PPEHWEEDAS WGPHRLAVLV PFRERFEELL VFVPHMRRFL SRKKIRHHIY VLNQVDHFRF NRAALINVGF LESSNSTDYI AMHDVDLLPL NEELDYGFPE AGPFHVASPE LHPLYHYKTY VGGILLLSKQ HYRLCNGMSN RFWGWGREDD EFYRRIKGAG LQLFRPSGIT TGYKTFRHLH DPAWRKRDQK RIAAQKQEQF KVDREGGLNT VKYHVASRTA LSVGGAPCTV LNIMLDCDKT ATPWCTFS (1-327 aa encoded by BC007317)
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,4-galactosyltransferase 7; beta-1,4-galactosyltransferase VII; Beta-1,4-GalTase 7; beta4Gal-T7; beta4GalT-VII; galactosyltransferase I; Proteoglycan UDP-galactose:beta-xylose beta1,4-galactosyltransferase I; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 7; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 7; UDP-galactose:beta-xylose beta-1,4-galactosyltransferase; XGalT-1; XGPT; Xylosylprotein 4-beta-galactosyltransferase; xylosylprotein beta 1,4-galactosyltransferase, polypeptide 7; Xylosylprotein beta-1,4-galactosyltransferase
Gene Aliases: B4GALT7; EDSP1; EDSSLA; UNQ748/PRO1478; XGALT1; XGPT1
UniProt ID: (Human) Q9UBV7
Entrez Gene ID: (Human) 11285
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support