Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
B7-H6 (NCR3LG1) is a transmembrane endogenous ligand expressed on the surfaces of tumor cells. B7-H6 binds with NKp30 present on NK cells. Binding of B7-H6 to NKp30 causes the ligation of NKp30 subsequently inducing NK cell activation and target cell cytolysis. B7-H6 is not constitutively expressed on normal tissue. Whether it can be expressed endogenously under certain conditions is under investigation. B7-H6 has been detected on circulating pro-inflammatory CD14(+)CD16(+) monocytes in some patients with sepsis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B7 homolog 6; B7-H6; DKFZp686I21167; Natural cytotoxicity triggering receptor 3 ligand 1; putative Ig-like domain-containing protein DKFZp686O24166/DKFZp686I21167
Gene Aliases: B7-H6; B7H6; DKFZp686O24166; NCR3LG1
UniProt ID: (Human) Q68D85
Entrez Gene ID: (Human) 374383
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support