Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KRSPSPSPT PEAKKKNAKK GPSTPYTKSK RGHREEEQED LTKDMDEPSP VPNVEEVTLP KTVNTKKDSE SAPVKGGTMT DLDEQEDESM ETTGKDEDEN STGNKGEQTK NPDLHEDNVT EQTHHIIIPS YAAWFDYNSV HAIERRALPE FFNGKNKSKT PEIYLAYRNF MIDTYRLNPQ EYLTSTACRR NLAGDVCAIM RVHAFLEQWG LINYQVDAES RPTPMGPPPT SHFHVLADTP SGLVPLQPKT PQGRQVDADT KAGRKGKELD DLVPETAKGK PELQTSASQQ MLNFPDKGKE KPTDMQNFGL RTDMYTKKNV PSKSKAAASA TREWTEQETL LLLEALEMY (300-647 aa encoded by BC013045)
Cdc73 is a tumor suppressor probably involved in transcriptional and post-transcriptional control pathways. Cdc73 is part of the PAF1 complex, a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9. PAF1 was first purified from yeast in association with RNA polymerase II and is believed to function in transcription elongation. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. In addition to coordinating events during transcription (initiation, promoter clearance, and elongation), hPAF also coordinates events in RNA quality control. Cdc73 may also be involved in cell cycle progression through the regulation of cyclin D1/PRAD1 expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAF170; BRG1-associated factor 170; chromatin remodeling complex BAF170 subunit; DKFZp313D0632; mammalian chromatin remodeling complex BRG1-associated factor 170; similar to SWI/SNF-related matrix-associated actin-dependent regulator of chromatin c2; SWI/SNF complex 170 kDa subunit; SWI/SNF complex subunit SMARCC2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 2; SWI3-like protein
Gene Aliases: 5930405J04Rik; BAF170; CRACC2; Rsc8; SMARCC2
UniProt ID: (Human) Q8TAQ2, (Mouse) Q6PDG5
Entrez Gene ID: (Rat) 685179, (Human) 6601, (Mouse) 68094
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support