Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 18
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form oligomers or heterodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. This protein also interacts with the tumor suppressor P53 after exposure to cell stress.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis regulator BAK; Bcl-2 homologous antagonist/killer; Bcl-2-like protein 7; Bcl2 homologous antagonist/killer; BCL2-antagonist/killer 1; Bcl2-L-7; BCL2-like 7 protein; BCL2-like 7protein; MGC117255; MGC3887; pro-apoptotic protein BAK
Gene Aliases: BAK; BAK-LIKE; BAK1; BCL2L7; CDN1; N-Bak; N-BAK1
UniProt ID: (Human) Q16611, (Mouse) O08734
Entrez Gene ID: (Human) 578, (Mouse) 12018, (Rat) 116502
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support