Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MDRHSSYIFI WLQLELCAMA VLLTKGEIRC YCDAAHCVAT GYMCKSELSA CFSRLLDPQN SNSPLTHGCL DSLASTTDIC QAKQARNHSG TTIPTLECCH EDMCNYRGLH DVLSPPRGEA SGQGNRYQHD GSRNLITKVQ ELTSSKELWF RAAVIAVPIA GGLTLVLLIM LALRMLRSEN KRLQDQRQQM LSRLHYSFHG HHSKKGQVAK LDLECMVPVS GHENCCLTCD KMRQADLSND KILSLVHWGM YSGHGKLEFV
BAMBI (BMP and activin membrane-bound inhibitor homolog, NMA) is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP and activin membrane-bound inhibitor homolog; Non-metastatic gene A protein; Putative transmembrane protein NMA
Gene Aliases: BAMBI; NMA
UniProt ID: (Human) Q13145
Entrez Gene ID: (Human) 25805
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support