Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: PDRLDLAQSL GLTQLQVKTW YQNRRMKWKK MVLKGGQEAP TKPKGRPKKN SIPTSEEIEA EEKMNSQAQG QEQLEPSQGQ EELCEAQEPK ARDVPLEMAE
Transcription factor. Binds optimally to the DNA consensus sequence 5'-YYTAATGRTTTTY-3'. May control the expression of neural adhesion molecules such as L1 or Ng-CAM during embryonic development of both the central and peripherical nervous system. May be involved in controlling adhesive processes in keratinizing epithelia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BarH-like homeobox 2; Homeobox protein BarH-like 2
Gene Aliases: BARX2
UniProt ID: (Human) Q9UMQ3
Entrez Gene ID: (Human) 8538
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support