Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSVLDALWED RDVRFDLSAQ QMKTRPGEVL IDCLDSIEDT KGNNGDRGRL LVTNLRILWH SLALSRVNVS VGYNCILNIT TRTANSKLRG QTEALYILTK CNSTRFEFIF TNLVPGSPRL FTSVMAVHRA YETSKMYRDF KLRSALIQNK QLRLLPQEHV YDKINGVWNL SSDQGNLGTF FITNVRIVWH ANMNDSFNVS IPYLQIRSIK IRDSKFGLAL VIESSQQSGG YVLGFKIDPV EKLQESVKEI NSLHKVYSAS PIFGVDYEME EKPQPLEALT VEQIQDDVEI DSDGHTDAFV AYFADGNKQQ DREPVFSEEL GLAIEKLKDG FTLQGLWEVM S
The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. The BBSome complex, together with the LTZL1, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. Required for BBSome complex ciliary localization but not for the proper complex assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bardet-Biedl syndrome 5 protein
Gene Aliases: BBS5
UniProt ID: (Human) Q8N3I7
Entrez Gene ID: (Human) 129880
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support