Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SEVQLMRLL PIKKSTALKV ALFCTTLSGD TSDTGEGTVV VSYGVSLENA VLDQLLNLGR QLISDHVDLV LCQKVIHPSL KQFLNMHRII AIDRIGVTLM EPLTKMTGTQ PIGSLGSICP NSYGSVKDVC TAKFGSKHFF HLIPNEATIC SLLLCNRNDT AWDELKLTCQ TALHVLQLTL KEPWALLGGG CTETHLAAYI RHKTHNDPES ILKDDECTQT ELQLIAEAFC SALESVVGSL EHDGGEILTD MKYGHLWSVQ ADSPCVANWP DLLSQCGCGL YNSQEELNWS FLRSTCRPFV PQSCLPHEAV VSASNLTLDC LTAKLSGLQV AVETANLILD LSYVIEDKN (223-570 aa encoded by BC028973)
Probable molecular chaperone. Assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. May play a role in protein processing in limb, cardiac and reproductive system development. May play a role in cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bardet-Biedl syndrome 6 protein; McKusick-Kaufman syndrome protein; McKusick-Kaufman syndrome protein; MKKS protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; Molecular chaperone MKKS; Protein Bbs6 homolog
Gene Aliases: 1300013E18Rik; AI463362; AI957237; BBS6; HMCS; KMS; MKKS; MKS
UniProt ID: (Human) Q9NPJ1, (Mouse) Q9JI70
Entrez Gene ID: (Human) 8195, (Mouse) 59030, (Rat) 311456
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support