Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDLILNRMD YLQVGVTSQK TMKLIPASRH RATQKVVIGD HDGVVMCFGM KKGEAAAVFK TLPGPKIARL ELGGVINTPQ EKIFIAAASE IRGFTKRGKQ FLSFETNLTE SIKAMHISGS DLFLSASYIY NHYCDCKDQH YYLSGDKIND VICLPVERLS RITPVLACQD RVLRVLQGSD VMYAVEVPGP PTVLALHNGN GGDSGEDLLF GTSDGKLALI QITTSKPVRK WEIQNEKKRG GILCIDSFDI VGDGVKDLLV GRDDGMVEVY SFDNANEPVL RFDQMLSESV TSIQGGCVGK DSYDEIVVST YSGWVTGLTT EPIHKESGPG EELKINQEMQ NKISSLRNEL E (1-350 aa encoded by BC032691)
BBS7, also named as BBS2L1, is a component of the BBSome complex which is required for ciliogenesis but is dispensable for centriolar satellite function. BBS7 is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bardet-Biedl syndrome 7 protein; Bardet-Biedl syndrome 7 protein homolog; BBS2-like 1; BBS2-like protein 1; FLJ10715
Gene Aliases: 8430406N16Rik; BBS2L1; BBS7
UniProt ID: (Human) Q8IWZ6, (Mouse) Q8K2G4
Entrez Gene ID: (Human) 55212, (Rat) 361930, (Mouse) 71492
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support