Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SILKDVLSKE ATKRKINLNI SYEINEVSVK HTLKLIHPKL EYQLLLAKKV QLIDALKELQ IHEGNTNFLI PEYHCILEEA DHLQEEYKKQ PAHLERLYG
BBS7, also named as BBS2L1, is a component of the BBSome complex which is required for ciliogenesis but is dispensable for centriolar satellite function. BBS7 is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bardet-Biedl syndrome 7 protein; BBS2-like 1; BBS2-like protein 1; FLJ10715
Gene Aliases: BBS2L1; BBS7
UniProt ID: (Human) Q8IWZ6
Entrez Gene ID: (Human) 55212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support