Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
Immunogen sequence: MNHLNVLAKA LYDNVAESPD ELSFRKGDIM TVLEQDTQGL DGWWLCSLHG RQGIVPGNRL KILVGMYDKK PAGPGSGPPA TPAQPQPGLH APAPPASQYT PMLPNTYQPQ PDSVYLVPTP SKAQQGLYQV PGPSPQFQSP PAKQTSTFSK QTPHHPFPNP ATDLYQVPPG PGGPAQDIYQ VPPSAGMGHD IYQVPPSMDT RSWEGTKPPA KVVVPTRVGQ GYVYEAAQPE QDEYDIPRHL LAPGPQDIYD VPPVRGLLPS QYGQEVYDTP PMAVKGPNGR DPLLEVYDVP PSVEKGLPPS NHHAVYDVPP SVSKDVPDGP LLREETYDVP PAFAKAKPFD PARTPLVLAA PPPDSPPAED VYDVPPPAPD LYDVPPGLRR PGPGTLYDVP RERVLPPEVA DGGVVDSGVY AVPPPAEREA
The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich regionp130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCA1; breast cancer anti-estrogen resistance 1; Breast cancer anti-estrogen resistance protein 1; Cas scaffolding protein family member 1; CRK-associated substrate; Crk-associated substrate p130Cas; OTTHUMP00000174941; p130cas; p130Cas (pTyr410); p130Cas (pY410); p130Cas phospho; phospho p130Cas
Gene Aliases: BCAR1; CAS; CAS1; CASS1; CRKAS; P130Cas
UniProt ID: (Human) P56945
Entrez Gene ID: (Human) 9564
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support