Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSRRKQGKPQ HLSKREFSPE PLEAILTDDE PDHGPLGAPE GDHDLLTCGQ CQMNFPLGDI LIFIEHKRKQ CNGSLCLEKA VDKPPSPS
This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell CLL/lymphoma 11A; B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2; B-cell lymphoma/leukemia 11A; BCL-11A; BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1; C2H2-type zinc finger protein; COUP-TF-interacting protein 1; ecotropic viral integration site 9 homolog; Ecotropic viral integration site 9 protein homolog; EVI-9; FLJ10173; FLJ34997; Zinc finger protein 856
Gene Aliases: BCL11A; BCL11A-L; BCL11a-M; BCL11A-S; BCL11A-XL; CTIP1; EVI9; HBFQTL5; KIAA1809; ZNF856
UniProt ID: (Human) Q9H165
Entrez Gene ID: (Human) 53335
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support