Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis facilitator Bcl-2-like protein 14; Apoptosis regulator Bcl-G; Bcl2-L-14; BCL2-like 14 (apoptosis facilitator); testicular tissue protein Li 26
Gene Aliases: BCL2L14; BCLG
UniProt ID: (Human) Q9BZR8
Entrez Gene ID: (Human) 79370
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support