Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SLYSGLSTPP ASYSMYSHLP VSSLLFSDEE FRDVRMPVAN PFPKERALPC DSARPVPGEY SRPTLEVSPN VCHSNIYSPK ETIPEEARSD MHYSVAEGLK PAAPSARNAP YFPCDKASKE EERPSSEDEI ALHFEPPNAP LNRKGLVSPQ SPQKSDCQPN SPTESCSSKN ACILQASGSP PAKSPTDPKA CNWKKYKFIV LNSLNQNAKP EGPEQAELGR LSPRAYTAPP ACQPPMEPEN LDLQSPTKLS ASGEDSTIPQ ASRLNNIVNR SMTGSPRSSS ESHSPLYMHP PKCTSCGSQS PQHAEMCLHT AGPTFPEEMG ETQSEYSDSS CENGAFFCNE CDCRFSEEAS
Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin's lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt's lymphomas and in nodular, lymphocyte predominant Hodgkin's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell lymphoma 5 protein; B-cell lymphoma 5 protein (BCL5); B-cell lymphoma 6 protein; B-cell lymphoma 6 protein transcript; BCL-5; BCL-6; cys his2 zinc finger transcription factor; cys-his2 zinc finger transcription factor; LAZ-3 protein; Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3); lymphoma-associated zinc finger gene on chromosome 3; Protein LAZ-3; Zinc finger and BTB domain-containing protein 27; Zinc finger and BTB domain-containing protein 27 (ZBTB27); Zinc finger protein 51; Zinc Finger Protein 51 (ZNF51); zinc finger transcription factor BCL6S
Gene Aliases: BCL5; BCL6; BCL6A; LAZ3; ZBTB27; ZNF51
UniProt ID: (Human) P41182
Entrez Gene ID: (Human) 604
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support