Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MGSPAAPEGA LGYVREFTRH SSDVLGNLNE LRLRGILTDV TLLVGGQPLR AHKAVLIACS GFFYSIFRGR AGVGVDVLSL PGGPEARGFA PLLDFMYTSR LRLSPATAPA VLAAATYLQM EHVVQACHRF IQASYEPLGI SLRPLEAEPP TPPTAPPPGS PRRSEGHPDP PTESRSCSQG PPSPASPDPK ACNWKKYKYI VLNSQASQAG SLVGERSSGQ PCPQARLPSG DEASSSSSSS SSSSSEEGPI PGPQSRLSPT AATVQFKCGA PASTPYLLTS QAQDTSGSPS ERARPLPGSE FFSCQNSEAV AGCSSGLDSL VPGDEDKPYK CQLCRSSFRY KGNLASHRTV HTGEKPYHCS ICGARFNRPA NLKTHSRIHS GEKPYKCETC GSRFVQVAHL RAHVLIHTGE KPYPCPTCGT RFRHLQTLKS HVRIHTGEKP YHCDPCGLHF RHKSQLRLHL RQKHGAATNT KVHYHILGGP
BCL6B acts as a sequence-specific transcriptional repressor in association with BCL6. BCL6B may function in a narrow stage or be related to some events in the early B-cell development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell CLL/lymphoma 6 member B protein; B-cell CLL/lymphoma 6, member B (zinc finger protein); Bcl6-associated zinc finger protein; Zinc finger protein 62
Gene Aliases: BAZF; BCL6B; ZBTB28; ZNF62
UniProt ID: (Human) Q8N143
Entrez Gene ID: (Human) 255877
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support