Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1. Involved in cell cycle progression, maintenance of the nuclear structure and stem cell differentiation. May play a role in lung tumor development or progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell CLL/lymphoma 7 protein family member B; B-cell CLL/lymphoma 7B
Gene Aliases: BCL7B
UniProt ID: (Human) C9JWD3
Entrez Gene ID: (Human) 9275
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support