Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLTKEEPVPEL LEAEAPEAYP VFEPVPPVPE AAQGDTEDSE GAPPLKRICP NAPD
Highest antigen sequence identity to the following orthologs - mouse 96%, rat 98%.
This gene is identified by the similarity of its product to the N-terminal region of BCL7A protein. The BCL7A protein is encoded by the gene known to be directly involved in a three-way gene translocation in a Burkitt lymphoma cell line. The function of this gene has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell CLL/lymphoma 7 protein family member C; B-cell CLL/lymphoma 7C
Gene Aliases: BCL7C
UniProt ID: (Human) Q8WUZ0
Entrez Gene ID: (Human) 9274
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support