Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total BCO2.
BCDO2 catalyzes the asymmetric oxidative cleavage of beta-carotene in carotene metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: b,b-carotene-9',10'-dioxygenase; B-diox-II; beta,beta-carotene 9',10'-dioxygenase variant 1; beta,beta-carotene 9',10'-dioxygenase variant 2; beta,beta-carotene 9',10'-dioxygenase variant 3; Beta,beta-carotene 9',10'-oxygenase; beta-carotene 9', 10'-dioxygenase 2; beta-carotene 9',10' oxygenase; beta-carotene 9',10'-dioxygenase 2; Beta-carotene dioxygenase 2; beta-diox-II; carotenoid 9',10' monooxygenase II; carotenoid 9',10' monoxygenase II; carotenoid-9',10'-cleaving dioxygenase; Carotenoid-cleaving dioxygenase, mitochondrial
Gene Aliases: Ab2-079; B-DIOX-II; BCDO2; Bcmo2; BCO2; CMO2
UniProt ID: (Human) Q9BYV7, (Mouse) Q99NF1
Entrez Gene ID: (Human) 83875, (Rat) 315644, (Mouse) 170752
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support