Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MLATRLSRPL SRLPGKTLSA CDRENGARRP LLLGSTSFIP IGRRTYASAA EPVGSKAVLV TGCDSGFGFS LAKHLHSKGF LVFAGCLMKD KGHDGVKELD SLNSDRLRTV QLNVCSSEEV EKVVEIVRSS LKDPEKGMWG LVNNAGISTF GEVEFTSLET YKQVAEVNLW GTVRMTKSFL PLIRRAKGRV VNISSMLGRM ANPARSPYCI TKFGVEAFSD CLRYEMYPLG VKVSVVEPGN FIAATSLYSP ESIQAIAKKM WEELPEVVRK DYGKKYFDEK IAKMETYCSS GSTDTSPVID AVTHALTATT PYTRYHPMDY YWWLRMQIMT HLPGAISDMI YIR
This gene encodes a member of the short-chain dehydrogenase/reductase gene family. The encoded protein forms a homotetrameric lipid-requiring enzyme of the mitochondrial membrane and has a specific requirement for phosphatidylcholine for optimal enzymatic activity. The encoded protein catalyzes the interconversion of acetoacetate and -3-hydroxybutyrate, the two major ketone bodies produced during fatty acid catabolism. Alternatively spliced transcript variants encoding the same protein have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (R)-3-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase (heart, mitochondrial); BDH; D-beta-hydroxybutyrate dehydrogenase, mitochondrial; MGC2723; MGC4347; MGC9788; Short chain dehydrogenase/reductase family 9C member 1; short chain dehydrogenase/reductase family 9C, member 1
Gene Aliases: BDH; BDH1; SDR9C1
UniProt ID: (Human) Q02338
Entrez Gene ID: (Human) 622
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support