Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGRLDGKVI ILTAAAQGIG QAAALAFARE GAKVIATDIN ESKLQELEKY PGIQTRVLDV TKKKQIDQFA NEVERLDVLF NVAGFVHHGT VLDCEEKDWD FSMNLNVRSM YLMIKAFLPK MLAQKSGNII NMSSVASSVK GVVNRCVYST TKAAVIGLTK SVAADFIQQG IRCNCVCPGT VDTPSLQERI QARGNPEEAR NDFLKRQKTG RFATAEEIAM LCVYLASDES TYVTGNPVII DGGWSL (1-245 aa encoded by B C095414)
Dehydrogenase that mediates the formation of 2,5-dihydroxybenzoic acid (2,5-DHBA), a siderophore that shares structural similarities with bacterial enterobactin and associates with LCN2, thereby playing a key role in iron assimilation and homeostasis. Plays a role in susceptibility to bacterial infection by providing an assimilable source of iron that is exploited by pathogenic bacteria. Also acts as a 3-hydroxybutyrate dehydrogenase (PubMed:16380372). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (R)-beta-hydroxybutyrate dehydrogenase; 3-hydroxybutyrate dehydrogenase type 2; 4-oxo-L-proline reductase; dehydrogenase/reductase (SDR family) member 6; Dehydrogenase/reductase SDR family member 6; FLJ13261; Oxidoreductase UCPA; R-beta-hydroxybutyrate dehydrogenase; Short chain dehydrogenase/reductase family 15C member 1; short chain dehydrogenase/reductase family 15C, member 1
Gene Aliases: BDH2; DHRS6; EFA6R; PRO20933; SDR15C1; UCPA-OR; UNQ6308; UNQ6308/PRO20933
UniProt ID: (Human) Q9BUT1
Entrez Gene ID: (Human) 56898
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support