Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ILALAEDKS DLENSVMQKK IKIPKLSLNH VEEDGEVKDY GEEDLQLRHI KRPEGRKPSE VAHKSIEAVV ARLEKQNGLS LGHSTCPEEV FVEASPGTED MDSLEDAVVP RALYEELLRN YQQQQEEMRH LQQELERTRR QLVQQAKKLK EYGALVSEMK ELRDLNRRLQ DVLLLRLGSG PAIDLEKVKS ECLEPEPELR STFSEEANTS SYYPAPAPVM DKYILDNGKV HLGSGIWVDE EKWHQLQVTQ GDSKYTKNLA VMIWGTDVLK NRSVTGVATK KKKDAVPKPP LSPHKLSIVR ECLYDRIAQE TVDETEIAQR LSKVNKYICE KIMDINKSCK NEERREAKYN LQ (71-421 aa encoded by BC007932)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEN domain-containing protein 5
Gene Aliases: 2310026E23Rik; BEND5; C1orf165
UniProt ID: (Human) Q7L4P6, (Mouse) Q8C6D4
Entrez Gene ID: (Human) 79656, (Mouse) 67621
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support