Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEN domain-containing protein 5
Gene Aliases: 2310026E23Rik; BEND5; C1orf165
UniProt ID: (Human) Q7L4P6, (Mouse) Q8C6D4
Entrez Gene ID: (Human) 79656, (Mouse) 67621, (Rat) 362564
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support