FIGURE: 1 / 2
This antibody is predicted to react with non-human primate based on sequence homology.
Signaling adapter molecule involved in p75NTR/NGFR signaling. Plays a role in cell cycle progression and neuronal differentiation. Inhibits neuronal differentiation in response to nerve growth factor (NGF). May act as a link between the cell cycle and neurotrophic factor signaling, possibly by functioning as an upstream modulator of receptor signaling, coordinating biological responses to external signals with internal cellular states.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain expressed, X-linked 1; Brain-expressed X-linked protein 1; H HGR74-h; ovarian granulosa cell 13.0 kDa protein hGR74; Protein BEX1
Gene Aliases: BEX1; BEX2; HBEX2; HGR74-h
UniProt ID: (Human) Q9HBH7
Entrez Gene ID: (Human) 55859
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support