Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGTKNKTRA QAKTEKKAAI QAKAGAEREA TGVVRPVAKT RAKAKAKTGS KTDAVAEMKA VSKNKVVAET KEGALSEPKT LGKAMGDFTP KAGNESTSST CKNEAGTDAW FWAGEEATIN SWFWNGEEAG NSFSTKNDKP EIGAQVCAEE LEPAAGADCK PRSGAEEEEE ENVIGNWFWE GDDTSFDPNP KPVSRIVKPQ PVYEINEKNR PKDWSEVTIW PNAPAVTPAV LGFRSQAPSE ASPPSYIVLA SAEENACSLP VATACRPSRN TRSCSQPIPE CRFDSDPCIQ TIDEIRRQIR IREVNGIKPF ACPCKMECYM DSEEFEKLVS LLKSTTDPLI HKIARIAMGV HNVHPFAQEF INEVGVVTLI ESLLSFPSPE MRKKTVITLN PPSGDERQRK IELHVKHMCK ETMSFPLNSP GQQSGLKILG QLTTDFVHHY IVANYFSELF HLLSSGNCKT RNLVLKLLLN MSENPTAARD MINMKALAAL KLIFNQKEAK ANLVSGVAIF INIKEHIRKG SIVVVDHLSY NTLMAIFREV KEIIETM
May play a role in the control of cellular aging and survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb9; G protein-coupled receptor associated sorting protein 3; p60-like protein; p60TRP; Protein BHLHb9; Transcription regulator of 60 kDa
Gene Aliases: BHLHB9; GASP3; GPRASP3; KIAA1701; p60TRP
UniProt ID: (Human) Q6PI77
Entrez Gene ID: (Human) 80823
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support