Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPPVGGKKAK KGILERLNAG EIVIGDGGFV FALEKRGYVK AGPWTPEAAV EHPEAVRQLH REFLRAGSNV MQTFTFYASE DKLENRGNYV LEKISGQEVN EAACDIARQV ADEGDALVAG GVSQTPSYLS CKSETEVKKV FLQQLEVFMK KNVDFLIAEY FEHVEEAVWA VETLIASGKP VAATMCIGPE GDLHGVPPGE CAVRLVKAGA SIIGVNCHFD PTISLKTVKL MKEGLEAARL KAHLMSQPLA YHTPDCNKQG FIDLPEFPFG LEPRVATRWD IQKYAREAYN LGVRYIGGCC GFEPYHIRAI AEELAPERGF LPPASEKHGS WGSGLDMHTK PWVRARARKE YWENLRIASG RPYNPSMSKP DGWGVTKGTA ELMQQKEATT EQQLKELFEK QKFKSQ
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Betaine--homocysteine S-methyltransferase 1; epididymis secretory sperm binding protein Li 61p
Gene Aliases: BHMT; BHMT1; HEL-S-61p
UniProt ID: (Human) Q93088
Entrez Gene ID: (Human) 635
Molecular Function:
methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support