Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: HL-60, Rat kidney; Cellular Location: Cytoplasm
Immunogen sequence: SGKPVAATMC IGPEGDLHGV PPGECAVRLV KAGASIIGVN CHFDPTISLK TVKLMKEGLE AARLKAHLMS QPLAYHTPDC NKQGFIDLPE FPFGLEPRVA TRWDIQKYAR EAYNLGVRYI GGCCGFEPYH IRAIAEELAP ERGFLPPASE KHGSWGSGLD MHTKPWVRAR ARKEYWENLR IASGRPYNPS MSKPDGWGVT KGTAELMQQK EATTEQQLKE LFEKQKFKSQ
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Betaine--homocysteine S-methyltransferase 1; betaine-homocysteine methyltransferase; epididymis secretory sperm binding protein Li 61p
Gene Aliases: BHMT; BHMT1; HEL-S-61p
UniProt ID: (Human) Q93088, (Rat) O09171
Entrez Gene ID: (Human) 635, (Rat) 81508
Molecular Function:
methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support