Invitrogen
Compatible with Direct ELISA
BHQ880 inhibits the activity of DKK1, a protein that negatively regulates the Wnt signaling pathway, thereby promoting bone formation and increasing bone density.
Protein Aliases: DKK1; UNQ492/PRO1008