Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GLLATLQDTQ KQLEHTRGSL SEQQEKVTRL TENLSALRRL QASKERQTAL DNEKDRDSHE DGDYYEVDIN GPEILACKYH VAVAEAGELR EQLKALRSTH EAREAQHAEE KGRYEAEGQA LTEKVSLLEK ASRQDRELLA RLEKELKKVS DVAGETQGSL SVA
BICD2 is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bic-D 2; bicaudal D homolog 2; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; Protein bicaudal D homolog 2; RP11-476B13.3
Gene Aliases: bA526D8.1; BICD2; KIAA0699; SMALED2
UniProt ID: (Human) O75181
Entrez Gene ID: (Human) 23299
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support