Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 10
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
The Bik protein is known to interact with cellular and viral survival-promoting proteins, such as BCL2 and the Epstein-Barr virus in order to enhance programmed cell death. Because its activity is suppressed in the presence of survival-promoting proteins, this protein is suggested as a likely target for antiapoptotic proteins. This protein shares a critical BH3 domain with other death-promoting proteins, BAX and BAK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis inducer NBK; apoptosis-inducing NBK; BBC1; Bcl-2-interacting killer; BCL2-interacting killer (apoptosis-inducing); bhikhari; Bik-like killer protein; BIP 1; BIP1; BP 4; BP4; cb 60; natural born killer
Gene Aliases: BIK; Biklk; BIP1; Blk; BP4; NBK
UniProt ID: (Human) Q13323, (Mouse) O70337
Entrez Gene ID: (Human) 638, (Mouse) 12124, (Rat) 114496
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support