Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31652 has been tested in indirect ELISA.
This gene encodes several isoforms of a nucleocytoplasmic adaptor protein, one of which was initially identified as a MYC-interacting protein with features of a tumor suppressor. Isoforms that are expressed in the central nervous system may be involved in synaptic vesicle endocytosis and may interact with dynamin, synaptojanin, endophilin, and clathrin. Isoforms that are expressed in muscle and ubiquitously expressed isoforms localize to the cytoplasm and nucleus and activate a caspase-independent apoptotic process. Studies in mouse suggest that this gene plays an important role in cardiac muscle development. Alternate splicing of the gene results in ten transcript variants encoding different isoforms. Aberrant splice variants expressed in tumor cell lines have also been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMPH 2; amphiphysin 2; Amphiphysin II; amphiphysin-like; Amphiphysin-like protein; box dependant MYC interacting protein 1; Box-dependent myc-interacting protein 1; Bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; Myc box-dependent-interacting protein 1; SH3 domain-containing protein 9
Gene Aliases: ALP-1; AMPH2; AMPHL; BIN1; BRAMP-2; SH3P9
UniProt ID: (Human) O00499, (Mouse) O08539
Entrez Gene ID: (Human) 274, (Mouse) 30948
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support