Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. The encoded protein inhibits apoptosis induced by serum deprivation but does not affect apoptosis resulting from exposure to menadione, a potent inducer of free radicals. The amino acid sequence predicts three baculovirus IAP repeat domains and a ring finger domain. Transcript variants encoding the same isoform have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIP 2; API2; Apoptosis inhibitor 2; baculoviral IAP repeat-containing 3; Baculoviral IAP repeat-containing protein 3; C-IAP2; Cellular inhibitor of apoptosis 2; H HIAP1; H MALT2; hIAP-1; IAP homolog; IAP homolog C; IAP-1; Inhibitor of apoptosis protein 1; mammalian IAP homolog C; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1
Gene Aliases: AIP1; API2; BIRC3; c-IAP2; CIAP2; HAIP1; HIAP1; MALT2; MIHC; RNF49
UniProt ID: (Human) Q13489
Entrez Gene ID: (Human) 330
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support