Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Biliverdin reductase B; biliverdin reductase B (flavin reductase (NADPH)); Biliverdin-IX beta-reductase; BVR-B; epididymis secretory protein Li 10; Flavin reductase (NADPH); FLR; FR; GHBP; Green heme-binding protein; NADPH; NADPH-dependent diaphorase; NADPH-flavin reductase; S-nitroso-CoA-assisted nitrosyltransferase; short chain dehydrogenase/reductase family 43U, member 1; SNO-CoA-assisted nitrosyltransferase
Gene Aliases: BLVRB; BVRB; FLR; HEL-S-10; SCAN; SDR43U1
UniProt ID: (Human) P30043
Entrez Gene ID: (Human) 645
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support