Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: MANVAENSSS DQRQACKKHE LYVSFRDLGW QDWIIAPEGY AAYYCEGECA FPLNSYMNAT NHAIVQTLVH FINPETVPKP CCAPTQLNAI SVLYFDDSSN VILKKYRNMV VRACGCH.
Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BMP-7. Anti-Human BMP-7-specific antibody was purified by affinity chromatography and then biotinylated.
Sandwich ELISA: To detect hBMP-7 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BMP-7 (500-P198) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBMP-7.
Western Blot: To detect Human BMP-7 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BMP-7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
500-P198BT-1MG will be provided as 2 x 500 µg
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP; BMP-7; Bone morphogenetic protein; Bone morphogenetic protein 7; OP-1; Osteogenic protein 1; RP11-560A15.5
Gene Aliases: BMP7; OP-1; OP1
UniProt ID: (Human) P18075
Entrez Gene ID: (Human) 655
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support