Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MHVRSLRAAA PHSFVALWAP LFLLRSALAD FSLDNEVHSS FIHRRLRSQE RREMQREILS ILGLPHRPRP HLQGKHNSAP MFMLDLYNAM AVEEGGGPGG QGFSYPYKAV FSTQGPPLAS LQDSHFLTDA DMVMSFVNLV EHDKEFFHPR YHHREFRFDL SKIPEGEAVT AAEFRIYKDY IRERFDNETF RISVYQVLQE HLGRESDLFL LDSRTLWASE EGWLVFDITA TSNHWVVNPR HNLGLQLSVE TLDGQSINPK LAGLIGRHGP QNKQPFMVAF FKATEVHFRS IRSTGSKQRS QNRSKTPKNQ EALRMANVAE NSSSDQRQAC KKHELYVSFR DLGWQDWIIA PEGYAAYYCE GECAFPLNSY MNATNHAIVQ TLVHFINPET VPKPCCAPTQ LNAISVLYFD DSSNVILKKY RNMVVRACGC H
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP; BMP-7; Bone morphogenetic protein; Bone morphogenetic protein 7; OP-1; Osteogenic protein 1; RP11-560A15.5
Gene Aliases: Bmp-7; BMP7; OP-1; OP1
UniProt ID: (Human) P18075, (Mouse) P23359
Entrez Gene ID: (Human) 655, (Mouse) 12162
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support