Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases which phosphorylates intracellular SMADs in response to bone morphogenic proteins. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activin A receptor, type II-like kinase 3; Activin receptor-like kinase 3; ALK-3; BMP type-1A receptor; BMP-2/BMP-4 receptor; BMPR-1A; BMPRIA; Bone morphogenetic protein 4 receptor; bone morphogenetic protein receptor type IA; Bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type 1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Gene Aliases: 10q23del; 1110037I22Rik; ACVRLK3; ALK3; AU045487; Bmpr; BMPR-IA; BMPR1A; CD292; SKR5
UniProt ID: (Human) P36894, (Rat) Q78EA7, (Mouse) P36895
Entrez Gene ID: (Human) 657, (Rat) 81507, (Mouse) 12166
Molecular Function:
serine/threonine protein kinase receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support