Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: AFPPPAAEAH PAARRGLRSP QLPSGAMSQN GAPGMQEESL QGSWVELHFS NNGNGGSVPA SVSIYNGDME KILLDAQHES G
Positive Samples: A375, Mouse heart; Cellular Location: Mitochondrion, Mitochondrion outer membrane, Single-pass membrane protein
NIP3 is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. NIP3 contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein is known to induce apoptosis, even in the presence of BCL2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCL2/adenovirus E1B 19 kDa protein-interacting protein 3; BCL2/adenovirus E1B 19 kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B 19 kDa-interacting protein 3; BCL2/adenovirus E1B 19kDa interacting protein 3; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 1, NIP3
Gene Aliases: BNIP3; NIP3
UniProt ID: (Human) Q12983, (Mouse) O55003
Entrez Gene ID: (Human) 664, (Rat) 84480, (Mouse) 12176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support