Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MASSNTVLMR LVASAYSIAQ KAGMIVRRVI AEGDLGIVEK TCATDLQTKA DRLAQMSICS SLARKFPKLT IIGEEDLPSE EVDQELIEDS QWEEILKQPC
BPNT1, also called bisphosphate 3-prime-nucleotidase, or BPntase, is a member of a magnesium-dependent phosphomonoesterase family. Lithium, a major drug used to treat manic depression, acts as an uncompetitive inhibitor of BPntase. The predicted human protein is 92% identical to mouse BPntase. BPntase's physiologic role in nucleotide metabolism may be regulated by inositol signaling pathways. The inhibition of human BPntase may account for lithium-induced nephrotoxicity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'(2'),5'-bisphosphate nucleotidase 1; 3'-phosphoadenosine 5'-phosphate phosphatase; Bisphosphate 3'-nucleotidase 1; BPntase; BPntase 1; epididymis luminal protein 20; HsPIP; Inositol-polyphosphate 1-phosphatase; PAP phosphatase; PAP-inositol 1,4-phosphatase; PAP-inositol-1,4-phosphatase; testicular tissue protein Li 29
Gene Aliases: BPNT1; HEL20; PIP
UniProt ID: (Human) O95861
Entrez Gene ID: (Human) 10380
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support