Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PDPSALQNL ASWNPSNPEC LLLVVKELVQ QYHQFQCSRL RESSRLMFEY QTLLEEPQYG ENMEIYAGKK NNWTGEFSAR FLLKLPVDFS NIPTYLLKDV NEDPGEDVAL LSVSFEDTEA TQVYPKLYLS PRIEHALGGS SALHIPAFPG GGCLIDYVPQ VCHLLTNKVQ YVIQGYHKRR EYIAAFLSHF GTGVVEYDAE GFTKLTLLLM WKDFCFLVHI DLPLFFPRDQ PTLTFQSVYH FTNSGQLYSQ AQKNYPYSPR WDGNEMAKRA KAYFKTFVPQ FQEAAFANGK L (94-383 aa encoded by BC001251)
BRCC45 was initially suggested to be a housekeeping protein that is highly expressed in brain and reproductive organs. Later experiments indicated BRCC45 forms a complex with the breast and ovarian predisposition proteins BRCA1 and BRCA2 as well as RAD51 and BRCC36. This complex has a ubiquitin E3 ligase activity and is thought to enhance cellular survival following DNA damage. BRCC45 has also been suggested to function as a death receptor-associated anti-apoptotic protein by inhibiting the BID-induced activation of the mitochondrial apoptotic pathway. Higher levels of BRCC45 were detected in the majority of hepatocellular carcinomas, suggesting that BRCC45 may promote tumorigenesis when overexpressed. At least three isoforms of BRCC45 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain and reproductive organ-expressed protein; BRCA1-A complex subunit BRE; BRCA1/BRCA2-containing complex subunit 45; BRCA1/BRCA2-containing complex, subunit 4; BRISC and BRCA1-A complex member 2
Gene Aliases: BABAM2; BRCC4; BRCC45; BRE
UniProt ID: (Human) Q9NXR7
Entrez Gene ID: (Human) 9577
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support