Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GSINDIKSD VCISSLVLKA NNIHSSPSFT HLDKSSPQKF LSNLSKEEIN LQRNIAGKVV TPHQKQAAGM SQETFEEKYR LSPTLSSTKG HLLIHSRPRS SSVKRKRVSH GSHSPPKEKC KRKRSTRRSI MPRLQLCRSE GRLQHVAGPA LEALSCGESS YDDYFSPDNL KERYSENLPP ESQLPSSPAQ LSCRSLSKKE RTSIFEMSDF SCVGKKTRTV DITNFTAKTI SSPRKTGNGE GRATSSCVTS APEEALRCCR QAGKEDACPE GNGFSYTIED PALPKGHDDD LTPLEGSLEE MKEAVGLKST QNKGTTSKIS NSSEGEAQSE HEPCFIVDCN METSTEEKEN LPGGYSGSM (253-610 aa encoded by BC030702)
This protein is implicated in chromosome condensation and DNA damage induced cellular responses, and may play a role in neurogenesis and regulation of the size of the cerebral cortex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRCT-repeat inhibitor of TERT expression 1; BRIT 1; FLJ12847; Microcephalin; truncated microcephalin
Gene Aliases: 5430437K10Rik; BRIT1; D030046N04Rik; MCPH1; MCT
UniProt ID: (Human) Q8NEM0, (Mouse) Q7TT79
Entrez Gene ID: (Human) 79648, (Mouse) 244329
Molecular Function:
DNA metabolism protein
damaged DNA-binding protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support