Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
BRP44L (brain protein 44-like protein), also known as HSPC040 or CGI-129, is a 109 amino acid mitochondrial protein belonging to the UPF0041 family. The gene that encodes BRP44L maps to human chromosome 6. Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis-regulating basic protein; Brain protein 44-like protein; HSPC040 protein; Mitochondrial pyruvate carrier 1
Gene Aliases: 0610006G08Rik; 3830411I18Rik; Arbp; BRP44L; CGI-129; dJ68L15.3; HSPC040; MPC1; MPYCD; PNAS-115
UniProt ID: (Human) Q9Y5U8, (Rat) P63031, (Mouse) P63030
Entrez Gene ID: (Human) 51660, (Rat) 171087, (Mouse) 55951
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support