Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to PAX5. It will not cross react with other PAX members.
Immunogen sequence: QPPNQPVPA SSHSIVSTGS VTQVSSVSTD SAGSSYSISG ILGITSPSAD TNKRKRDEGI QESPVPNGHS LPGRDFLRKQ MRGDLFTQQQ LEVLDRVFER QHYSDIFTTT EPIKPEQTTE YSAMASLAGG LDDMKANLAS PTPADIGSSV PGPQSYPIVT GRDLASTTLP GYPPHVPPAG QGSYSAPTLT GMVPGSEFSG SPYSHPQYSS YNDSWRFPNP GLLGSPYYYS AAARGAAPPA AATAYDRH (145-392 aa encoded by BC156927)
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; paired box homeotic gene 5; Paired box protein Pax-5; paired domain gene 5; transcription factor PAX 5
Gene Aliases: ALL3; BSAP; PAX5
UniProt ID: (Human) Q02548
Entrez Gene ID: (Human) 5079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support