Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: WGGIWPRHRF SLQVNIRKRD NSRKEVQRRI SAHQPGPEGQ EESTPQSDVT EDGESPEDPS GTEGQLSEEE KPDQQPLSGE EELEPEASDG SGSWEDAAL
This gene encodes protein seipin, which is located in the endoplasmic reticulum and may be important for lipid droplet morphology. Mutations in this gene have been associated with congenital generalized lipodystrophy type 2 or Berardinelli-Seip syndrome, a rare autosomal recessive disease characterized by a near absence of adipose tissue and severe insulin resistance. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Berardinelli-Seip congenital lipodystrophy 2 (seipin); Bernardinelli-Seip congenital lipodystrophy type 2 protein; Seipin
Gene Aliases: BSCL2; GNG3LG; HMN5; PELD; SPG17
UniProt ID: (Human) Q96G97
Entrez Gene ID: (Human) 26580
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support