Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Bone marrow stromal cell antigen 1 (BST1) is a stromal cell line-derived glycosylphosphatidylinositol-anchored molecule that facilitates pre-B cell growth. BST1, which is also known as ADP-rybosyl cyclase 2 and DAMP-1, is an enzyme which is expressed by BST1 gene in human. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis. The polyclonal B cell abnormalities in rheumatoid arthritis may be, at least in part, attributed to BST1 overexpression in the stromal cell population.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; bone marrow stromal antigen 1; Bone marrow stromal cell antigen 1; BST-1; cADPR hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; NAD(+) nucleosidase; NAD+ nucleosidase
Gene Aliases: BST1; CD157
UniProt ID: (Human) Q10588
Entrez Gene ID: (Human) 683
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support