Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
BTG1 is a member of an anti-proliferative gene family that regulates cell growth and differentiation. Expression of this gene is highest in the G0/G1 phases of the cell cycle and downregulated when cells progressed through G1. The encoded protein interacts with several nuclear receptors, and functions as a coactivator of cell differentiation. This locus has been shown to be involved in a t(8; 12)(q24; q22) chromosomal translocation in a case of B-cell chronic lymphocytic leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell translocation gene 1 protein; B-cell translocation gene 1, anti-proliferative; Protein BTG1
Gene Aliases: APRO2; BTG1
UniProt ID: (Human) P62324
Entrez Gene ID: (Human) 694
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support