Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKMASSLAFL LLNFHVSLFL VQLLTPCSAQ FSVLGPSGPI LAMVGEDADL PCHLFPTMSA ETMELRWVSS SLRQVVNVYA DGKEVEDRQS APYRGRTSIL RDGITAGKAA LRIHNVTASD SGKYLCYFQD GDFYEKALVE LKVAALGSDL HIEVKGYEDG GIHLECRSTG WYPQPQIKWS DTKGENIPAV EAPVVADGVG LYAVAASVIM RGSSGGGVSC IIRNSLLGLE KTASISIADP FFRSAQPWIA ALAGTLPISL LLLAGASYFL WRQQKEKIAL SRETEREREM KEMGYAATEQ EISLREKLQE ELKWRKIQYM ARGEKSLAYH EWKMALFKPA DVILDPDTAN AILLVSEDQR SVQRAEEPRD LPDNPERFEW RYCVLGCENF TSGRHYWEVE VGDRKEWHIG VCSKNVERKK GWVKMTPENG YWTMGLTDGN KYRALTEPRT NLKLPEPPRK VGIFLDYETG EISFYNATDG SHIYTFPHAS FSEPLYPVFR ILTLEPTALT ICPIPKEVES SPDPDLVPDH SLETPLTPGL ANESGEPQAE VTSLLLPAHP GAEVSPSATT NQNHKLQART EALY
Plays a role in T-cell responses in the adaptive immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: butyrophilin 3; Butyrophilin subfamily 3 member A3; butyrophilin subfamily 3 member A3 secreted isoform; butyrophilin, subfamily 3, member A3
Gene Aliases: BTF3; BTN3.3; BTN3A3
UniProt ID: (Human) O00478
Entrez Gene ID: (Human) 10384
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support