Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ISIADPFFRS AQPWIAALAG TLPISLLLLA GASYFLWRQQ KEKIALSRET EREREMKEMG YAATEQEISL REKLQEELKW RKIQYMARGE KSLAYHEWKM ALFKPADVIL DPDTANAILL VSEDQRSVQR AEEPRDLPDN PERFEWRYCV LGCENFTSGR HYWEVEVGDR KEWHIGVCSK NVERKKGWVK MTPENGYWTM GLTDGNKYRA LTEPRTNLKL PEPPRKVGIF LDYETGEISF YNATDGSHIY TFPHASFSEP LYPVFRILTL EPTALTICPI PKEVESSPDP DLVPDHSLET PLTPGLANES GEPQAEVTSL LLPAHPGAEV SPSATTNQNH KLQARTEALY
Plays a role in T-cell responses in the adaptive immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: butyrophilin 3; Butyrophilin subfamily 3 member A3; butyrophilin subfamily 3 member A3 secreted isoform; butyrophilin, subfamily 3, member A3
Gene Aliases: BTF3; BTN3.3; BTN3A3
UniProt ID: (Human) O00478
Entrez Gene ID: (Human) 10384
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support