Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDPAEAVLQE KALKFMCSMP RSLWLGCSSL ADSMPSLRCL YNPGTGALTA FQNSSEREDC NNGEPPRKII PEKNSLRQTY NSCARLCLNQ ETVCLASTAM KTENCVAKTK LANGTSSMIV PKQRKLSASY EKEKELCVKY FEQWSESDQV EFVEHLISQM CHYQHGHINS YLKPMLQRDF ITALPARGLD HIAENILSYL DAKSLCAAEL VCKEWYRVTS DGMLWKKLIE RMVRTDSLWR GLAERRGWGQ YLFKNKPPDG NAPPNSFYRA LYPKIIQDIE TIESNWRCGR HSLQRIHCRS ETSKGVYCLQ YDDQKIVSGL RDNTIKIWDK NTLECKRILT GHTGSVLCLQ YDERVIITGS SDSTVRVWDV NTGEMLNTLI HHCEAVLHLR FNNGMMVTCS KDRSIAVWDM ASPTDITLRR VLVGHRAAVN VVDFDDKYIV SASGDRTIKV WNTSTCEFVR TLNGHKRGIA CLQYRDRLVV SGSSDNTIRL WDIECGACLR VLEGHEELVR CIRFDNKRIV SGAYDGKIKV WDLVAALDPR APAGTLCLRT LVEHSGRVFR LQFDEFQIVS SSHDDTILIW DFLNDPAAQA EPPRSPSRTY TYISR
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbws class; in addition to an F-box, this protein contains multiple WD-40 repeats. This protein is homologous to Xenopus bTrCP1, yeast Met30, Neurospora Scon2 and Drosophila Slimb proteins. It interacts with HIV-1 Vpu and connects CD4 to the proteolytic machinery. It also associates specifically with phosphorylated IkappaBalpha and beta-catenin destruction motifs, probably functioning in multiple transcriptional programs by activating the NF-kappaB pathway and inhibiting the beta-catenin pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-TrCP1; E3RSIkappaB; Epididymis tissue protein Li 2a; F-box and WD repeats protein beta-TrCP; F-box and WD-repeat protein 1B; F-box/WD repeat-containing protein 1A; pIkappaBalpha-E3 receptor subunit
Gene Aliases: BETA-TRCP; betaTrCP; BTRC; BTRCP; bTrCP1; FBW1A; FBXW1; FBXW1A; FWD1
UniProt ID: (Human) Q9Y297
Entrez Gene ID: (Human) 8945
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support