Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KLVYVHHLL GEGAFAQVYE ATQGDLNDAK NKQKFVLKVQ KPANPWEFYI GTQLMERLKP SMQHMFMKFY SAHLFQNGSV LVGELYSYGT LLNAINLYKN TPEKVMPQGL VISFAMRMLY MIEQVHDCEI IHGDIKPDNF ILGNGFLEQD DEDDLSAGLA LIDLGQSIDM KLFPKGTIFT AKCETSGFQC VEMLSNKPWN YQIDYFGVAA TVYCMLFGTY MKVKNEGGEC KPEGLFRRLP HLDMWNEFFH VMLNIPDCHH LPSLDLLRQK LKKVFQQHYT NKIRALRNRL IVLLLECKRS (785-1083 aa encoded by BC028201)
BUB1A is a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BUB1 budding uninhibited by benzimidazoles 1 homolog; BUB1, mitotic checkpoint serine/threonine kinase; BUB1A; budding uninhibited by benzimidazoles 1 homolog; hBUB1; Mitotic checkpoint serine/threonine-protein kinase BUB1; mitotic spindle checkpoint kinase; putative serine/threonine-protein kinase
Gene Aliases: BUB1; BUB1A; BUB1L; hBUB1
UniProt ID: (Human) O43683
Entrez Gene ID: (Human) 699, (Dog) 100682599
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support