Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Batoclimab (HBM9161) is a fully human antibody targeting the neonatal Fc receptor (FcRn). It targets patients with neuromyelitis optica spectrum diseases and other autoimmune diseases, and can accelerate the degradation of autoantibodies in their bodies. Batoclimab (HBM9161) is an antibody that targets the neonatal Fc receptor (FcRn). FcRn expression is up-regulated by pro-inflammatory cytokines (such as TNF-alpha) and prolongs the half-life of IgG and serum albumin by reducing lysosomal degradation in endothelial cells and bone marrow-derived cells. Blocking the FcRn-IgG interaction can accelerate the degradation of autoantibodies and reduce the onset of various pathogenic IgG-mediated autoimmune diseases, including myasthenia gravis, thyroid ophthalmopathy, neuromyelitis optica spectrum disease and immunity Thrombocytopenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HL161
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support