Search Thermo Fisher Scientific
PA1-75031 specifically detects Benzimidazole.
PA1-75031 has been successfully used in ELISA applications.
The PA1-75031 immunogen is 2-Amino-5-(propylthio)benzimidazole-BTG.
Benzimidazole is a heterocyclic aromatic organic compound. This class includes anthelmintic drugs such as albendazole, mebendazole and triclabendazole.
Protein Aliases: 2-Amino-5-(propylthio)-benzimidazole; C7H6N2